Product Details
- SNP ID
-
rs202044123
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:64978440 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCAGTATGGATTATCTTATGTGT[A/T]TTAAGGGCTGAGGACTGGTTAAAGG
- Phenotype
-
MIM: 194624
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF117
PubMed Links
Gene Details
- Gene
- ZNF117
- Gene Name
- zinc finger protein 117
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015852.3 |
2416 |
Missense Mutation |
AAA,AAT |
K377N |
NP_056936.2 |
View Full Product Details