Product Details

SNP ID
rs201787553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:22727528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCCCAGTACCCCCAGGAGAAG[A/T]TTCCAAAGATGTAGCCGCCCCACAC
Phenotype
MIM: 147620
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IL6 PubMed Links

Gene Details

Gene
IL6
Gene Name
interleukin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000600.4 558 Missense Mutation GAT,GTT D35V NP_000591.1
NM_001318095.1 558 Intron NP_001305024.1
XM_005249745.4 558 Missense Mutation GAT,GTT D89V XP_005249802.1
XM_011515390.2 558 Missense Mutation GAT,GTT D35V XP_011513692.1
Gene
LOC541472
Gene Name
uncharacterized LOC541472
There are no transcripts associated with this gene.

View Full Product Details