Product Details
- SNP ID
-
rs201978453
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:64979232 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCATGCAAAATGTTTTGCGACA[C/T]TCTTTACATTTAAAATGTTTATTTT
- Phenotype
-
MIM: 194624
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF117
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3807068] are located under a probe and SNP(s) [rs3807069] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF117
- Gene Name
- zinc finger protein 117
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015852.3 |
1624 |
Silent Mutation |
|
|
NP_056936.2 |
View Full Product Details