Product Details

SNP ID
rs199648871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102466154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCTTCACCCTCTCTTCCCCAG[C/G]TCAATGACAACCTGAAAGTCTCCTT
Phenotype
MIM: 613302 MIM: 615167 MIM: 610929 MIM: 604150
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
There are no transcripts associated with this gene.

Gene
LRWD1
Gene Name
leucine rich repeats and WD repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317721.1 532 UTR 5 NP_001304650.1
NM_152892.2 532 Missense Mutation CTC,GTC L106V NP_690852.1
XM_017011845.1 532 UTR 5 XP_016867334.1
Gene
MIR4467
Gene Name
microRNA 4467
There are no transcripts associated with this gene.

Gene
MIR5090
Gene Name
microRNA 5090
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
There are no transcripts associated with this gene.

Gene
POLR2J
Gene Name
RNA polymerase II subunit J
There are no transcripts associated with this gene.

View Full Product Details