Product Details

SNP ID
rs199713239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124486003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGTCAGCTTTTGGAATTGAGCT[G/T]CTTCCTCGAAAAGGTCCCTCGCTGT
Phenotype
MIM: 603046
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF139 PubMed Links

Gene Details

Gene
RNF139
Gene Name
ring finger protein 139
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007218.3 697 Silent Mutation CTG,CTT L118L NP_009149.2
XM_017013002.1 697 UTR 5 XP_016868491.1
Gene
TATDN1
Gene Name
TatD DNase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details