Product Details

SNP ID
rs201313143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:68780596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCGATTTCATTCCTGTGTCACC[A/G]GCACCGTCACCCACTCGGGGAATTG
Phenotype
MIM: 602745
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM122A PubMed Links

Gene Details

Gene
FAM122A
Gene Name
family with sequence similarity 122A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138333.4 591 Silent Mutation CCA,CCG P186P NP_612206.4
Gene
PIP5K1B
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278253.1 591 Intron NP_001265182.1
NM_003558.3 591 Intron NP_003549.1
XM_005252261.3 591 Intron XP_005252318.1
XM_005252262.4 591 Intron XP_005252319.1
XM_006717300.2 591 Intron XP_006717363.1
XM_006717301.1 591 Intron XP_006717364.1
XM_011519082.2 591 Intron XP_011517384.1
XM_011519083.2 591 Intron XP_011517385.1
XM_011519084.2 591 Intron XP_011517386.1
XM_017015188.1 591 Intron XP_016870677.1
XM_017015189.1 591 Intron XP_016870678.1
XM_017015190.1 591 Intron XP_016870679.1
XM_017015191.1 591 Intron XP_016870680.1

View Full Product Details