Product Details

SNP ID
rs199968559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13318932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAAATGGTGTCGACAGCGGCCT[A/G]TACTGTGCCTTCACTGATGTCATCA
Phenotype
MIM: 300920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN3L PubMed Links

Gene Details

Gene
ATXN3L
Gene Name
ataxin 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135995.1 1468 Nonsense Mutation CAG,TAG Q335* NP_001129467.1
Gene
GS1-600G8.3
Gene Name
unknown transcript
There are no transcripts associated with this gene.

View Full Product Details