Product Details

SNP ID
rs202122673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154532204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCACCTGCCATAAATATAGG[A/G]GATGGGCTTGGGCTTCTCCAGCTCA
Phenotype
MIM: 305900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G6PD PubMed Links

Gene Details

Gene
G6PD
Gene Name
glucose-6-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000402.4 1568 Missense Mutation CCC,TCC P511S NP_000393.4
NM_001042351.2 1568 Missense Mutation CCC,TCC P481S NP_001035810.1

View Full Product Details