Product Details

SNP ID
rs201960773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13319237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGTTTCTTGGCGGCTTAGTTCA[A/G]GGGCCCTCTGAAAATCCTCCTCATC
Phenotype
MIM: 300920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN3L PubMed Links

Gene Details

Gene
ATXN3L
Gene Name
ataxin 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135995.1 1163 Missense Mutation CCT,CTT P233L NP_001129467.1
Gene
GS1-600G8.3
Gene Name
unknown transcript
There are no transcripts associated with this gene.

View Full Product Details