Product Details

SNP ID
rs559077429
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41654798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATGATATAGTGCTTGAAGACAT[A/G]GAAAACCTCTTCTCTATGGACATGC
Phenotype
MIM: 612641 MIM: 610772
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANK1 PubMed Links

Gene Details

Gene
ANK1
Gene Name
ankyrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000037.3 6761 UTR 3 NP_000028.3
NM_001142445.1 6761 UTR 3 NP_001135917.1
NM_001142446.1 6761 UTR 3 NP_001135918.1
NM_020475.2 6761 UTR 3 NP_065208.2
NM_020476.2 6761 UTR 3 NP_065209.2
NM_020477.2 6761 UTR 3 NP_065210.2
NM_020478.4 6761 UTR 3 NP_065211.2
NM_020480.4 6761 UTR 3 NP_065213.2
XM_005273476.4 6761 UTR 3 XP_005273533.1
XM_011544490.2 6761 UTR 3 XP_011542792.1
XM_011544491.2 6761 UTR 3 XP_011542793.1
XM_011544494.2 6761 UTR 3 XP_011542796.1
XM_011544495.2 6761 UTR 3 XP_011542797.1
XM_011544496.2 6761 UTR 3 XP_011542798.1
XM_011544500.2 6761 UTR 3 XP_011542802.1
XM_011544501.2 6761 UTR 3 XP_011542803.1
XM_011544502.2 6761 UTR 3 XP_011542804.1
XM_011544503.2 6761 UTR 3 XP_011542805.1
XM_011544504.2 6761 UTR 3 XP_011542806.1
XM_011544505.2 6761 UTR 3 XP_011542807.1
XM_017013319.1 6761 UTR 3 XP_016868808.1
XM_017013320.1 6761 UTR 3 XP_016868809.1
XM_017013321.1 6761 UTR 3 XP_016868810.1
XM_017013322.1 6761 UTR 3 XP_016868811.1
XM_017013323.1 6761 UTR 3 XP_016868812.1
XM_017013324.1 6761 UTR 3 XP_016868813.1
XM_017013325.1 6761 UTR 3 XP_016868814.1
XM_017013326.1 6761 UTR 3 XP_016868815.1
XM_017013327.1 6761 UTR 3 XP_016868816.1
XM_017013328.1 6761 UTR 3 XP_016868817.1
XM_017013329.1 6761 UTR 3 XP_016868818.1
Gene
MIR486-1
Gene Name
microRNA 486-1
There are no transcripts associated with this gene.

Gene
MIR486-2
Gene Name
microRNA 486-2
There are no transcripts associated with this gene.

Gene
NKX6-3
Gene Name
NK6 homeobox 3
There are no transcripts associated with this gene.

View Full Product Details