Product Details

SNP ID
rs530895640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37779725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACACGAAGCTCCTTGGTCTTTAT[C/T]AACAAATAGAGTTCTATAATCTCTT
Phenotype
MIM: 606489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC3 PubMed Links

Gene Details

Gene
EXOSC3
Gene Name
exosome component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002269.2 1830 UTR 3 NP_001002269.1
NM_016042.3 1830 UTR 3 NP_057126.2
Gene
LOC105376037
Gene Name
uncharacterized LOC105376037
There are no transcripts associated with this gene.

Gene
TRMT10B
Gene Name
tRNA methyltransferase 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286950.1 1830 Intron NP_001273879.1
NM_001286951.1 1830 Intron NP_001273880.1
NM_001286952.1 1830 Intron NP_001273881.1
NM_001286953.1 1830 Intron NP_001273882.1
NM_001286954.1 1830 Intron NP_001273883.1
NM_144964.3 1830 Intron NP_659401.2
XM_005251375.2 1830 UTR 3 XP_005251432.1
XM_005251379.2 1830 UTR 3 XP_005251436.1
XM_011517735.2 1830 UTR 3 XP_011516037.1
XM_011517736.2 1830 UTR 3 XP_011516038.1
XM_011517738.2 1830 UTR 3 XP_011516040.1
XM_011517739.2 1830 UTR 3 XP_011516041.1
XM_011517742.1 1830 Intron XP_011516044.1
XM_011517743.2 1830 Intron XP_011516045.1
XM_017014313.1 1830 UTR 3 XP_016869802.1

View Full Product Details