Product Details

SNP ID
rs375728200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136496290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGTAGCTGTGCTGCGAGGGGGG[C/T]GTCAGGAACTGGGCTGCGGTCACGG
Phenotype
MIM: 190198
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOTCH1 PubMed Links

Gene Details

Gene
NOTCH1
Gene Name
notch 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017617.4 6741 Silent Mutation ACA,ACG T2483T NP_060087.3
XM_011518717.2 6741 Silent Mutation ACA,ACG T2242T XP_011517019.2

View Full Product Details