Product Details

SNP ID
rs537424617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59831763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGATATTGTTGATAGAAGAGAC[G/A]ACAAGGCCCCAAGATGTCATTGTGG
Phenotype
MIM: 609342
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 1141 Silent Mutation GTC,GTT V369V NP_005133.2
XM_011544939.2 1141 Silent Mutation GTC,GTT V355V XP_011543241.1

View Full Product Details