Product Details

SNP ID
rs398122377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53939261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCCACCTCGTCCAGCGCCACC[C/-]TGGGCTACGGCTACCCCTTCGGGGG
Phenotype
MIM: 142976
Polymorphism
C/-, Insertion/deletion
Allele Nomenclature
Literature Links
HOXC13 PubMed Links

Gene Details

Gene
HOXC13
Gene Name
homeobox C13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017410.2 470 Frame Shift Delete CTG,TGG L119W NP_059106.2
Gene
HOXC13-AS
Gene Name
HOXC13 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details