Product Details

SNP ID
rs367543053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23412855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCAGATCAAGATGTGGCAAAGC[T/C]ACTCCTCATTCAAGCCCTTTTGAAA
Phenotype
MIM: 616096 MIM: 613613 MIM: 160710 MIM: 160760
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MHRT PubMed Links

Gene Details

Gene
MHRT
Gene Name
myosin heavy chain-associated RNA transcript
There are no transcripts associated with this gene.

Gene
MIR208B
Gene Name
microRNA 208b
There are no transcripts associated with this gene.

Gene
MYH6
Gene Name
myosin heavy chain 6
There are no transcripts associated with this gene.

Gene
MYH7
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
There are no transcripts associated with this gene.

View Full Product Details