Product Details

SNP ID
rs567224368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28842605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGATAGAACATTCTTTCCCACT[A/C]TCTCCCAGGAGGCTGAATAACGGCA
Phenotype
MIM: 607931 MIM: 608937 MIM: 602389
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATXN2L PubMed Links

Gene Details

Gene
ATXN2L
Gene Name
ataxin 2 like
There are no transcripts associated with this gene.

Gene
MIR4721
Gene Name
microRNA 4721
There are no transcripts associated with this gene.

Gene
SH2B1
Gene Name
SH2B adaptor protein 1
There are no transcripts associated with this gene.

Gene
TUFM
Gene Name
Tu translation elongation factor, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003321.4 1877 UTR 3 NP_003312.3
XM_011545928.2 1877 Intron XP_011544230.1
XM_017023619.1 1877 Intron XP_016879108.1

View Full Product Details