Product Details

SNP ID
rs587776428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23603515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTGTCCAGCCAGCAAATGAG[AG/-]TCTGTACCCGACCATTTCACAAAAG
Phenotype
MIM: 603836 MIM: 610355
Polymorphism
AG/-, Insertion/deletion
Allele Nomenclature
Literature Links
NDUFAB1 PubMed Links

Gene Details

Gene
NDUFAB1
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
There are no transcripts associated with this gene.

Gene
PALB2
Gene Name
partner and localizer of BRCA2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024675.3 4237 Frame Shift Delete GACTCT,GATCCA DS1168DP NP_078951.2
XM_011545946.2 4237 Frame Shift Delete GACTCT,GATCCA DS1170DP XP_011544248.1
XM_011545947.2 4237 UTR 3 XP_011544249.1
XM_011545948.2 4237 Frame Shift Delete GACTCT,GATCCA DS873DP XP_011544250.1
XM_017023671.1 4237 Frame Shift Delete GACTCT,GATCCA DS1091DP XP_016879160.1
XM_017023672.1 4237 Frame Shift Delete GACTCT,GATCCA DS1089DP XP_016879161.1
XM_017023673.1 4237 UTR 3 XP_016879162.1

View Full Product Details