Product Details

SNP ID
rs763513849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23603495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATACAAATATATTTCCATCTTT[T/C]TGTCCAGCCAGCAAATGAGAGTCTG
Phenotype
MIM: 603836 MIM: 610355
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
NDUFAB1 PubMed Links

Gene Details

Gene
NDUFAB1
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
There are no transcripts associated with this gene.

Gene
PALB2
Gene Name
partner and localizer of BRCA2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024675.3 4258 Silent Mutation CAA,CAG Q1175Q NP_078951.2
XM_011545946.2 4258 Silent Mutation CAA,CAG Q1177Q XP_011544248.1
XM_011545947.2 4258 UTR 3 XP_011544249.1
XM_011545948.2 4258 Silent Mutation CAA,CAG Q880Q XP_011544250.1
XM_017023671.1 4258 Silent Mutation CAA,CAG Q1098Q XP_016879160.1
XM_017023672.1 4258 Silent Mutation CAA,CAG Q1096Q XP_016879161.1
XM_017023673.1 4258 UTR 3 XP_016879162.1

View Full Product Details