Product Details

SNP ID
rs372149439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63872486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGTTCTCTTGGGTCAGCGCCT[G/T]ACTGCTACAAAGAGGGCAGCAGTGT
Phenotype
MIM: 118820 MIM: 139240 MIM: 612756
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSH2 PubMed Links

Gene Details

Gene
CSH2
Gene Name
chorionic somatomammotropin hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020991.3 Intron NP_066271.1
NM_022644.3 Intron NP_072170.1
NM_022645.2 Intron NP_072171.1
Gene
GH2
Gene Name
growth hormone 2
There are no transcripts associated with this gene.

Gene
TCAM1P
Gene Name
testicular cell adhesion molecule 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details