Product Details

SNP ID
rs572425083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41813078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCACAGCCTCCTCCGGCTCC[C/G]CCTGCTGCAGTTGCTGCTACTGGTG
Phenotype
MIM: 607063
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FKBP10 PubMed Links

Gene Details

Gene
FKBP10
Gene Name
FK506 binding protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021939.3 369 Missense Mutation CCC,CGC P15R NP_068758.3
XM_011525099.2 369 Missense Mutation CCC,CGC P15R XP_011523401.1
XM_011525100.2 369 UTR 5 XP_011523402.1
Gene
P3H4
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006455.2 369 Intron NP_006446.1
XM_006721640.3 369 Intron XP_006721703.1

View Full Product Details