Product Details

SNP ID
rs765314515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47941766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCACCTGTGTGGTCGCAGTGCT[G/C]CCATGGACCTGGGACCCATGCGCAA
Phenotype
MIM: 603287 MIM: 601801
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
PNPO PubMed Links

Gene Details

Gene
PNPO
Gene Name
pyridoxamine 5'-phosphate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018129.3 196 Missense Mutation CCC,GCC P31A NP_060599.1
XM_005257500.3 196 UTR 5 XP_005257557.1
XM_011524968.2 196 UTR 5 XP_011523270.1
XM_017024813.1 196 Intron XP_016880302.1
Gene
SP2
Gene Name
Sp2 transcription factor
There are no transcripts associated with this gene.

Gene
SP2-AS1
Gene Name
SP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details