Product Details

SNP ID
rs774176392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15999805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCTCGGCGCGTGGTCGGGGGCC[A/G]GGAGCGCGTCTGGCCTGCAGTGCGC
Phenotype
MIM: 613814 MIM: 614535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC19 PubMed Links

Gene Details

Gene
TTC19
Gene Name
tetratricopeptide repeat domain 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271420.1 426 UTR 5 NP_001258349.1
NM_017775.3 426 UTR 5 NP_060245.3
XM_017024801.1 426 Missense Mutation CAG,CGG Q107R XP_016880290.1
XM_017024802.1 426 Missense Mutation CAG,CGG Q107R XP_016880291.1
Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 426 Intron NP_001036162.1
NM_001042698.1 426 Intron NP_001036163.1

View Full Product Details