Product Details

SNP ID
rs777064041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53882326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCGGAGCCGGCGCGCCCGGGGT[G/C]CCGCTCCCTGCCTGGCGCGCTCCGC
Phenotype
MIM: 609959 MIM: 176980
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
MYADM PubMed Links

Gene Details

Gene
MYADM
Gene Name
myeloid associated differentiation marker
There are no transcripts associated with this gene.

Gene
PRKCG
Gene Name
protein kinase C gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316329.1 143 UTR 5 NP_001303258.1
NM_002739.4 143 UTR 5 NP_002730.1

View Full Product Details