Product Details
- SNP ID
-
rs587782684
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:28687962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGCACCCTCGGCTTCCCCTTCAC[G/-]GGGCCGCTTTCGACTAGTAGAAGGC
- Phenotype
-
MIM: 604373
MIM: 615098
- Polymorphism
- G/-, Insertion/deletion
- Allele Nomenclature
-
- Literature Links
-
CHEK2
PubMed Links
Gene Details
- Gene
- CHEK2
- Gene Name
- checkpoint kinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005735.1 |
1459 |
Frame Shift Delete |
CGT,GTG |
R566V |
NP_001005735.1 |
NM_001257387.1 |
1459 |
Frame Shift Delete |
CGT,GTG |
R302V |
NP_001244316.1 |
NM_007194.3 |
1459 |
Frame Shift Delete |
CGT,GTG |
R523V |
NP_009125.1 |
NM_145862.2 |
1459 |
Frame Shift Delete |
CGT,GTG |
R494V |
NP_665861.1 |
XM_006724114.3 |
1459 |
Frame Shift Delete |
CGT,GTG |
R374V |
XP_006724177.2 |
XM_006724116.2 |
1459 |
Frame Shift Delete |
CGT,GTG |
R342V |
XP_006724179.2 |
XM_011529839.2 |
1459 |
Frame Shift Delete |
CGT,GTG |
R576V |
XP_011528141.1 |
XM_011529840.2 |
1459 |
Intron |
|
|
XP_011528142.1 |
XM_011529841.1 |
1459 |
Frame Shift Delete |
CGT,GTG |
R499V |
XP_011528143.1 |
XM_011529842.2 |
1459 |
Frame Shift Delete |
CGT,GTG |
R466V |
XP_011528144.1 |
XM_011529843.1 |
1459 |
Frame Shift Delete |
CGT,GTG |
R456V |
XP_011528145.1 |
XM_011529844.2 |
1459 |
Intron |
|
|
XP_011528146.1 |
XM_011529845.2 |
1459 |
Frame Shift Delete |
CGT,GTG |
R302V |
XP_011528147.1 |
XM_017028560.1 |
1459 |
Frame Shift Delete |
CGT,GTG |
R564V |
XP_016884049.1 |
XM_017028561.1 |
1459 |
Frame Shift Delete |
CGT,GTG |
R302V |
XP_016884050.1 |
- Gene
- TTC28
- Gene Name
- tetratricopeptide repeat domain 28
There are no transcripts associated with this gene.
View Full Product Details