Product Details

SNP ID
rs398123548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154532154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCTCACCCCGCCCCTGCCCGC[T/C]GGGCTCTGTCCCCAGCCCCCACCCT
Phenotype
MIM: 305900
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
G6PD PubMed Links

Gene Details

Gene
G6PD
Gene Name
glucose-6-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000402.4 Intron NP_000393.4
NM_001042351.2 Intron NP_001035810.1

View Full Product Details