Product Details

SNP ID
rs587780526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46500372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATAGGGTTTCTCTCCTGTATGA[A/G]TTCTCTGATGAACAGAGAGGTGTGA
Phenotype
MIM: 300573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 1615 Missense Mutation ACT,ATT T406I NP_001034980.1
NM_001146291.1 1615 Missense Mutation ACT,ATT T400I NP_001139763.1
NM_001190417.1 1615 Missense Mutation ACT,ATT T401I NP_001177346.1
XM_011543941.2 1615 Intron XP_011542243.1
XM_011543943.2 1615 Intron XP_011542245.1
XM_011543944.2 1615 Intron XP_011542246.1
XM_011543945.2 1615 Intron XP_011542247.1
XM_017029728.1 1615 Intron XP_016885217.1
XM_017029729.1 1615 Intron XP_016885218.1
XM_017029730.1 1615 Missense Mutation ACT,ATT T339I XP_016885219.1
XM_017029731.1 1615 Intron XP_016885220.1
XM_017029732.1 1615 Intron XP_016885221.1
XM_017029733.1 1615 Intron XP_016885222.1

View Full Product Details