Product Details

SNP ID
rs778073001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46500598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTTTATAAAATGCTTGTCTAA[TG/-]TGTGTTTTTTCATGTCTAATAAGAT
Phenotype
MIM: 300573
Polymorphism
TG/-, Insertion/deletion
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 1388 Frame Shift Delete CAATAG,CACATT Q*330HI NP_001034980.1
NM_001146291.1 1388 Frame Shift Delete CAATAG,CACATT Q*324HI NP_001139763.1
NM_001190417.1 1388 Frame Shift Delete CAATAG,CACATT Q*325HI NP_001177346.1
XM_011543941.2 1388 Intron XP_011542243.1
XM_011543943.2 1388 Intron XP_011542245.1
XM_011543944.2 1388 Intron XP_011542246.1
XM_011543945.2 1388 Intron XP_011542247.1
XM_017029728.1 1388 Intron XP_016885217.1
XM_017029729.1 1388 Intron XP_016885218.1
XM_017029730.1 1388 Frame Shift Delete CAATAG,CACATT Q*263HI XP_016885219.1
XM_017029731.1 1388 Intron XP_016885220.1
XM_017029732.1 1388 Intron XP_016885221.1
XM_017029733.1 1388 Intron XP_016885222.1

View Full Product Details