Product Details

SNP ID
rs782764609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154531480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTATAGCAGAGAGGCTGCCTA[C/T]GGGTGGCACGGGGTGGCCATGGAGT
Phenotype
MIM: 305900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
G6PD PubMed Links

Gene Details

Gene
G6PD
Gene Name
glucose-6-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000402.4 2195 UTR 3 NP_000393.4
NM_001042351.2 2195 UTR 3 NP_001035810.1

View Full Product Details