Product Details

SNP ID
rs374625049
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:186444306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAATACCATCTTCATAAATGTG[AAC/-]AACAATTGTGGTGATCTTCAGTTC
Phenotype
MIM: 171490
Polymorphism
AAC/-, Insertion/deletion
Allele Nomenclature
Literature Links
LOC102724919 PubMed Links

Gene Details

Gene
LOC102724919
Gene Name
uncharacterized LOC102724919
There are no transcripts associated with this gene.

Gene
PDC
Gene Name
phosducin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002597.4 486 Silent Mutation NP_002588.3
NM_022576.3 486 Silent Mutation NP_072098.1
XM_011509603.2 486 Frame Shift Delete GTT,N/A V138N/A XP_011507905.1

View Full Product Details