Product Details

SNP ID
rs528653413
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:11787652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTTCGGAATGTTTCAGCTGTT[C/G]TGAGAAAACACTTTGCATTCCTGCC
Phenotype
MIM: 607093
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf167 PubMed Links

Gene Details

Gene
C1orf167
Gene Name
chromosome 1 open reading frame 167
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010881.1 5228 Intron NP_001010881.1
XM_011541269.2 5228 Intron XP_011539571.2
XM_011541271.2 5228 Intron XP_011539573.2
XM_011541272.2 5228 Intron XP_011539574.1
XM_011541274.2 5228 Intron XP_011539576.2
XM_011541275.2 5228 Intron XP_011539577.2
XM_011541276.2 5228 Intron XP_011539578.1
XM_011541277.2 5228 Intron XP_011539579.1
XM_011541278.2 5228 Intron XP_011539580.1
XM_011541280.2 5228 Intron XP_011539582.2
XM_011541281.2 5228 Intron XP_011539583.2
XM_017001062.1 5228 Intron XP_016856551.1
XM_017001063.1 5228 Intron XP_016856552.1
XM_017001064.1 5228 Intron XP_016856553.1
Gene
LOC102724659
Gene Name
uncharacterized LOC102724659
There are no transcripts associated with this gene.

Gene
MTHFR
Gene Name
methylenetetrahydrofolate reductase (NAD(P)H)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005957.4 5228 UTR 3 NP_005948.3
XM_005263458.3 5228 Intron XP_005263515.1
XM_005263460.4 5228 Intron XP_005263517.1
XM_005263462.4 5228 Intron XP_005263519.1
XM_005263463.3 5228 Intron XP_005263520.1
XM_011541495.2 5228 Intron XP_011539797.1
XM_011541496.2 5228 Intron XP_011539798.1
XM_017001328.1 5228 Intron XP_016856817.1

View Full Product Details