Product Details

SNP ID
rs537891059
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:197084179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATAAAACTATAAATGATAAAA[A/C]TGAAGAATGTAATGAACAGTTTATG
Phenotype
MIM: 605481
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 10836 UTR 3 NP_001193775.1
NM_018136.4 10836 UTR 3 NP_060606.3

View Full Product Details