Product Details

SNP ID
rs760528024
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:43403223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGTTAATGAAAAAGGATTATC[G/A]AATCTCCCGAAATGTTCGCCTGGCT
Phenotype
MIM: 615463
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SZT2 PubMed Links

Gene Details

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 158 Missense Mutation CAA,CGA Q25R NP_056099.3
XM_005270686.3 158 Missense Mutation CAA,CGA Q46R XP_005270743.1
XM_006710501.3 158 Missense Mutation CAA,CGA Q25R XP_006710564.1
XM_011541106.2 158 Missense Mutation CAA,CGA Q6R XP_011539408.1
XM_011541107.2 158 Intron XP_011539409.1
XM_017000819.1 158 Missense Mutation CAA,CGA Q46R XP_016856308.1
XM_017000820.1 158 Missense Mutation CAA,CGA Q46R XP_016856309.1
XM_017000821.1 158 Intron XP_016856310.1

View Full Product Details