Product Details

SNP ID
rs772662249
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43403241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTATCGAATCTCCCGAAATGTTC[G/A]CCTGGCTTGGTTCCTCAGTCATCTG
Phenotype
MIM: 615463
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SZT2 PubMed Links

Gene Details

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 176 Missense Mutation CAC,CGC H31R NP_056099.3
XM_005270686.3 176 Missense Mutation CAC,CGC H52R XP_005270743.1
XM_006710501.3 176 Missense Mutation CAC,CGC H31R XP_006710564.1
XM_011541106.2 176 Missense Mutation CAC,CGC H12R XP_011539408.1
XM_011541107.2 176 Intron XP_011539409.1
XM_017000819.1 176 Missense Mutation CAC,CGC H52R XP_016856308.1
XM_017000820.1 176 Missense Mutation CAC,CGC H52R XP_016856309.1
XM_017000821.1 176 Intron XP_016856310.1

View Full Product Details