Product Details

SNP ID
rs539573652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177392809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCCATGGCGGAGGCGGCGGCTG[C/T]AGCGGGTGGGACTGGCTTGGGCGCG
Phenotype
MIM: 603051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGPS PubMed Links

Gene Details

Gene
AGPS
Gene Name
alkylglycerone phosphate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003659.3 67 Missense Mutation GCA,GTA A7V NP_003650.1
XM_011512041.2 67 Intron XP_011510343.1
XM_011512042.1 67 Intron XP_011510344.1
XM_011512043.2 67 Intron XP_011510345.1
Gene
LOC100130691
Gene Name
uncharacterized LOC100130691
There are no transcripts associated with this gene.

View Full Product Details