Product Details

SNP ID
rs730880027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219250479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCATGCCCTCACCCACATAC[C/T]AGTGCACAAACGCCCTCTTGGCATA
Phenotype
MIM: 604719 MIM: 191110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLB1L PubMed Links

Gene Details

Gene
GLB1L
Gene Name
galactosidase beta 1 like
There are no transcripts associated with this gene.

Gene
STK16
Gene Name
serine/threonine kinase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011512067.2 1055 Intron XP_011510369.1
XM_017005160.1 1055 Intron XP_016860649.1
Gene
TUBA4A
Gene Name
tubulin alpha 4a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278552.1 1055 Nonsense Mutation TAG,TGG *392W NP_001265481.1
NM_006000.2 1055 Nonsense Mutation TAG,TGG *407W NP_005991.1
XM_017004824.1 1055 Nonsense Mutation TAG,TGG *254W XP_016860313.1
Gene
TUBA4B
Gene Name
tubulin alpha 4b
There are no transcripts associated with this gene.

View Full Product Details