Product Details

SNP ID
rs780650245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171785296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCACTGAGTGGCTGCCACTGCT[G/-]CCTTTGGCTGAACCACAGCAACACT
Phenotype
MIM: 603667
Polymorphism
G/-, Insertion/deletion
Allele Nomenclature
Literature Links
LOC105373738 PubMed Links

Gene Details

Gene
LOC105373738
Gene Name
uncharacterized LOC105373738
There are no transcripts associated with this gene.

Gene
SLC25A12
Gene Name
solute carrier family 25 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003705.4 2179 Frame Shift Delete GAG,GCA E672A NP_003696.2
XM_011512070.2 2179 Frame Shift Delete GAG,GCA E581A XP_011510372.1

View Full Product Details