Product Details

SNP ID
rs866957815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206765617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCTTCTCGGGGAAGCTGTCAT[G/A]GCTGCTCCTGTACGTAGTCACGGTC
Phenotype
MIM: 612322
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
FASTKD2 PubMed Links

Gene Details

Gene
FASTKD2
Gene Name
FAST kinase domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136193.1 230 UTR 5 NP_001129665.1
NM_001136194.1 230 UTR 5 NP_001129666.1
NM_014929.3 230 UTR 5 NP_055744.2
Gene
MDH1B
Gene Name
malate dehydrogenase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039845.2 230 Intron NP_001034934.1
NM_001282940.1 230 Intron NP_001269869.1
XM_005246315.4 230 Intron XP_005246372.1
XM_005246316.4 230 Intron XP_005246373.1
XM_011510628.2 230 Intron XP_011508930.1
XM_017003373.1 230 Intron XP_016858862.1
XM_017003374.1 230 Intron XP_016858863.1

View Full Product Details