Product Details

SNP ID
rs377468133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108908142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACATTGGAGAAGTCGAAGCTCTT[G/A]TAAACAATCTCCAGGAGATCCTGAT
Phenotype
MIM: 605128
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005459.3 578 Silent Mutation TAC,TAT Y170Y NP_005450.3
XM_005247910.3 578 Nonsense Mutation CAA,TAA Q184* XP_005247967.1
XM_011513334.2 578 Silent Mutation TAC,TAT Y86Y XP_011511636.1

View Full Product Details