Product Details

SNP ID
rs371346159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70198086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGAGAGGCCAGTTTTGCCCAA[G/T]GAGCCCAGGCAGGCCAAGTTGATCC
Phenotype
MIM: 614843
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ODAM PubMed Links

Gene Details

Gene
ODAM
Gene Name
odontogenic, ameloblast asssociated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017855.3 321 Nonsense Mutation GGA,TGA G102* NP_060325.3
XM_005265698.3 321 Nonsense Mutation GGA,TGA G119* XP_005265755.1
XM_006714245.3 321 Nonsense Mutation GGA,TGA G119* XP_006714308.1
XM_017008331.1 321 Nonsense Mutation GGA,TGA G88* XP_016863820.1

View Full Product Details