Product Details

SNP ID
rs370051225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43446398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAACCATGACGAATCCCATCTC[G/A]GATATGATGTGAAGTCCAGGATATA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TMEM267 PubMed Links

Gene Details

Gene
TMEM267
Gene Name
transmembrane protein 267
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022483.4 1855 Nonsense Mutation CGA,TGA R158* NP_071928.2
XM_005248333.4 1855 Nonsense Mutation CGA,TGA R158* XP_005248390.1
XM_005248335.3 1855 Nonsense Mutation CGA,TGA R158* XP_005248392.1
XM_006714486.3 1855 Nonsense Mutation CGA,TGA R158* XP_006714549.1
XM_011514074.2 1855 Nonsense Mutation CGA,TGA R158* XP_011512376.1
XM_011514075.2 1855 Nonsense Mutation CGA,TGA R158* XP_011512377.1

View Full Product Details