Product Details

SNP ID
rs567470870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92199851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAATGGGTCTCTGAAATCTTCA[T/A]TTTTAAGAGGCATCTAGGTAATTAT
Phenotype
MIM: 604214
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
KRIT1 PubMed Links

Gene Details

Gene
KRIT1
Gene Name
KRIT1, ankyrin repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013406.1 4115 UTR 3 NP_001013424.1
NM_004912.3 4115 UTR 3 NP_004903.2
NM_194454.1 4115 UTR 3 NP_919436.1
NM_194455.1 4115 UTR 3 NP_919437.1
NM_194456.1 4115 UTR 3 NP_919438.1
XM_005250660.3 4115 Intron XP_005250717.1
XM_005250662.3 4115 Intron XP_005250719.1
XM_005250665.3 4115 Intron XP_005250722.1
XM_005250666.3 4115 Intron XP_005250723.1
XM_005250667.2 4115 UTR 3 XP_005250724.1
XM_005250668.3 4115 Intron XP_005250725.1
XM_005250669.3 4115 Intron XP_005250726.1
XM_006716161.3 4115 Intron XP_006716224.1
XM_006716162.3 4115 Intron XP_006716225.1
XM_006716163.3 4115 Intron XP_006716226.1
XM_011516651.2 4115 Intron XP_011514953.1
XM_011516653.2 4115 UTR 3 XP_011514955.1
XM_011516654.2 4115 Intron XP_011514956.1
XM_011516655.2 4115 Intron XP_011514957.1
XM_011516656.2 4115 Intron XP_011514958.1
XM_011516657.2 4115 Intron XP_011514959.1
XM_011516658.2 4115 Intron XP_011514960.1
XM_011516659.2 4115 Intron XP_011514961.1
XM_011516660.2 4115 Intron XP_011514962.1
XM_011516661.2 4115 Intron XP_011514963.1
XM_017012755.1 4115 UTR 3 XP_016868244.1
XM_017012756.1 4115 Intron XP_016868245.1
XM_017012757.1 4115 Intron XP_016868246.1
XM_017012758.1 4115 Intron XP_016868247.1
XM_017012759.1 4115 Intron XP_016868248.1
Gene
MIR1285-1
Gene Name
microRNA 1285-1
There are no transcripts associated with this gene.

View Full Product Details