Product Details

SNP ID
rs769212821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:146116732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCGGAAGAACTGGAGCCTGGAG[G/C]GGGGTGAGGGGAGAAGAGGAAGCGG
Phenotype
MIM: 604569
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
CNTNAP2 PubMed Links

Gene Details

Gene
CNTNAP2
Gene Name
contactin associated protein-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014141.5 698 UTR 5 NP_054860.1
XM_017011950.1 698 UTR 5 XP_016867439.1

View Full Product Details