Product Details

SNP ID
rs770717871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:50463316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACATGGGCAGATTCCACCGTG[C/T]GAGAACAGATGGCAAAGCGCAGGAC
Phenotype
MIM: 107930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDC PubMed Links

Gene Details

Gene
DDC
Gene Name
dopa decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000790.3 1330 Missense Mutation NP_000781.1
NM_001082971.1 1330 Missense Mutation NP_001076440.1
NM_001242886.1 1330 Missense Mutation NP_001229815.1
NM_001242887.1 1330 Missense Mutation NP_001229816.1
NM_001242888.1 1330 Missense Mutation NP_001229817.1
NM_001242889.1 1330 Missense Mutation NP_001229818.1
NM_001242890.1 1330 Intron NP_001229819.1
XM_005271745.4 1330 Missense Mutation XP_005271802.1
XM_011515161.2 1330 Missense Mutation XP_011513463.2

View Full Product Details