Product Details

SNP ID
rs1830834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149967586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAATTATTTATATCTAGCAGTATC[C/T]GGGTTGAGTTTCCATTTCTTTTGGA
Phenotype
MIM: 605697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ULBP1 PubMed Links

Gene Details

Gene
ULBP1
Gene Name
UL16 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317089.1 Intron NP_001304018.1
NM_025218.3 Intron NP_079494.1
XM_017011322.1 Intron XP_016866811.1

View Full Product Details