Product Details

SNP ID
rs35518197
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:25122994 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGATGCAGAATCTGACAATCTTTA[C/T]CTTTATAGGTTGGGAGTTTTGGGCA
Phenotype
MIM: 610286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCMT1 PubMed Links

Gene Details

Gene
LCMT1
Gene Name
leucine carboxyl methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032391.1 Intron NP_001027563.1
NM_016309.2 Intron NP_057393.2
XM_005255354.3 Intron XP_005255411.1
XM_011545862.2 Intron XP_011544164.1
XM_011545864.1 Intron XP_011544166.1

View Full Product Details