Product Details
- SNP ID
-
rs10540
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:494662 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCCAGGGTTGCCTCGAGGCCGGTC[A/G]TCCAGGGAGAGCAGCAGCAGGAAGA
- Phenotype
-
MIM: 612793
MIM: 173320
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PTDSS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77508999] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PTDSS2
- Gene Name
- phosphatidylserine synthase 2
There are no transcripts associated with this gene.
- Gene
- RNH1
- Gene Name
- ribonuclease/angiogenin inhibitor 1
View Full Product Details