Product Details
- SNP ID
-
rs12582133
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:93681852 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATGTTTTTGAAAATCCTGTATACC[A/G]TAAGTTCTAGAGGTGTGTGTGTGTG
- Phenotype
-
MIM: 603454
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRADD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79528197] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRADD
- Gene Name
- CASP2 and RIPK1 domain containing adaptor with death domain
View Full Product Details