Product Details

SNP ID
rs13339
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:34610705 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATAGGGAGGTCCACTTTTGTGGACT[C/T]AAACCTTGATGGGGATGTTGAGCAG
Phenotype
MIM: 607387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCTN3 PubMed Links

Gene Details

Gene
DCTN3
Gene Name
dynactin subunit 3
There are no transcripts associated with this gene.

Gene
RPP25L
Gene Name
ribonuclease P/MRP subunit p25 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148178.2 882 UTR 3 NP_680544.1
NM_148179.2 882 UTR 3 NP_680545.1

View Full Product Details