Product Details

SNP ID
rs15706
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64230768 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAACAACGGGCCAGGCCCCTTC[T/C]CTCTGCCCCCGGGTGCTTGAAGTCT
Phenotype
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links
Additional Information
For this assay, SNP(s) [rs80058187] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307980.1 337 UTR 5 NP_001294909.1
NM_001307981.1 337 UTR 5 NP_001294910.1
NM_005528.3 337 UTR 5 NP_005519.2
Gene
FERMT3
Gene Name
fermitin family member 3
There are no transcripts associated with this gene.

Gene
NUDT22
Gene Name
nudix hydrolase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128612.2 337 Intron NP_001122084.1
NM_001128613.2 337 Intron NP_001122085.1
NM_001271831.1 337 Intron NP_001258760.1
NM_032344.3 337 Intron NP_115720.1
Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
There are no transcripts associated with this gene.

Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details