Product Details

SNP ID
rs2235982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33413027 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTCTAACCATATGACCTCGGAC[A/G]AGTCCCTTAACCTCTAGGAGCCTCA
Phenotype
MIM: 616953 MIM: 603763 MIM: 602881 MIM: 603384
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUTA PubMed Links

Gene Details

Gene
CUTA
Gene Name
cutA divalent cation tolerance homolog (E. coli)
There are no transcripts associated with this gene.

Gene
KIFC1
Gene Name
kinesin family member C1
There are no transcripts associated with this gene.

Gene
PHF1
Gene Name
PHD finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002636.4 Intron NP_002627.1
NM_024165.2 Intron NP_077084.1
XM_011514662.1 Intron XP_011512964.1
XM_011514663.1 Intron XP_011512965.1
XM_011514664.1 Intron XP_011512966.1
XM_011514665.1 Intron XP_011512967.1
XM_011514666.1 Intron XP_011512968.1
XM_011514669.1 Intron XP_011512971.1
XM_011514670.2 Intron XP_011512972.1
XM_017010939.1 Intron XP_016866428.1
Gene
SYNGAP1
Gene Name
synaptic Ras GTPase activating protein 1
There are no transcripts associated with this gene.

View Full Product Details